Alimemazine Tartrate ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate, Alimemazine Hemitartrate

A151143-100MG A151143-1G A151143-5G
ALA151143100MGEA 134.81 USD
ALA151143100MG ALA151143-1G ALA151143-5G
Alimemazine Tartrate ≥98% (by HPLC, titration analysis)
Alimemazine Tartrate
Formula: C₃₆H₄₄N₄S₂·C₄H₆O₆
Melting Pt: 153 - 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00242594
CAS Number: 4330-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd6b833fb45bc331da2268be3abae278e0bbe096&vid=bd6b833fb45bc331da2268be3abae278e0bbe096&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR