5-Allyl-3-methoxysalicylaldehyde ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

A151139-1G A151139-250MG
ALA151139-1GEA 43.61 USD
ALA151139-1G ALA151139250MG
5-Allyl-3-methoxysalicylaldehyde ≥98%
5-Allyl-3-methoxysalicylaldehyde
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 167°C
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD06408022
CAS Number: 22934-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8013aacfa11549693e64c6007e70fab6a788232e&vid=8013aacfa11549693e64c6007e70fab6a788232e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR