5-Amino-1H-tetrazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1H-Tetrazol-5-ylamine

A151135-100G A151135-25G
ALA151135-100GEA 84.46 USD
ALA151135-100G ALA151135-25G
5-Amino-1H-tetrazole ≥98%
5-Amino-1H-tetrazole
Formula: CH₃N₅
MW: 85.07 g/mol
Melting Pt: 201 °C
MDL Number: MFCD00005248
CAS Number: 4418-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b149d4cb40dfe2a96fec1076a8e0a55557bbf365&vid=b149d4cb40dfe2a96fec1076a8e0a55557bbf365&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR