2-Amino-6-iodopurine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151133-1G A151133-25G A151133-5G
ALA151133-1GEA 17.96 USD
ALA151133-1G ALA151133-25G ALA151133-5G
2-Amino-6-iodopurine ≥98%
2-Amino-6-iodopurine
Formula: C₅H₄IN₅
Melting Pt: 240°C
Storage Temperature: Ambient
MDL Number: MFCD08706375
CAS Number: 19690-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6875bd057456d537fabc54aac6bc509b0d2fb68&vid=d6875bd057456d537fabc54aac6bc509b0d2fb68&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR