2-Azido-1,3-dimethylimidazolinium hexafluorophosphate ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

A151129-1G A151129-5G
ALA151129-1GEA 150.96 USD
ALA151129-1G ALA151129-5G
2-Azido-1,3-dimethylimidazolinium hexafluorophosphate ≥98% (by HPLC)
2-Azido-1,3-dimethylimidazolinium hexafluorophosphate

Efficient diazo transfer reagent for the preparation of azides from primary amines. An alternative to tosyl azide with an easier-to-separate water soluble by-product。

Formula: C₅H₁₀F₆N₅P
Storage Temperature: Freezer
MDL Number: MFCD18782891
CAS Number: 1266134-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR