Pentyl nitrite (mixed isomers ) ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151126-100ML A151126-25ML
ALA151126100MLEA 183.26 USD
ALA151126100ML ALA151126-25ML
Pentyl nitrite (mixed isomers ) ≥95% (by GC)
Pentyl nitrite (mixed isomers )
Formula: C₅H₁₁NO₂
Boiling Pt: 104 °C
Density: 0.883 g/cm³
Flash Pt: -18°C
Storage Temperature: Refrigerator
MDL Number: MFCD00036132
CAS Number: 463-04-7
UN: 1113
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ecd68795db9f22d8ec887ece0e8cbb88b3a6cde&vid=5ecd68795db9f22d8ec887ece0e8cbb88b3a6cde&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR