2-(2-Aminophenyl)ethanol ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-(2-Aminophenyl)ethyl alcohol, 2-Aminophenethyl alcohol

A151124-100G A151124-1G A151124-25G A151124-5G
ALA151124-100GEA 399.86 USD
ALA151124-100G ALA151124-1G ALA151124-25G ALA151124-5G
2-(2-Aminophenyl)ethanol ≥95% (by GC)
2-(2-Aminophenyl)ethanol

2-Aminophenethyl alcohol undergoes one-pot cyclization with carboxylic acids in the presence of PPh3, CCl4 and NEt3 to yield N-acyl indolines.2-Aminophenethyl alcohol was used in the synthesis of: 1. indole derivatives. 2. N-(cyanothioformyl)indoline. 3. dihydro-3,1-benzoxazepine.

Formula: C₈H₁₁NO
Boiling Pt: 147-148 °C
Density: 1.12 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007754
CAS Number: 5339-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fca5dfd96fa7f73a73bfcada5c0ccce2b2b47d6&vid=1fca5dfd96fa7f73a73bfcada5c0ccce2b2b47d6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR