2-Amino-1-phenylethanol ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
alpha-(Aminomethyl)benzyl Alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine
A151123-1G
A151123-25G
A151123-5G
ALA151123-1GEA
46.46
USD
ALA151123-1G
ALA151123-25G
ALA151123-5G
2-Amino-1-phenylethanol ≥97% (by GC)
2-Amino-1-phenylethanol
Formula:
C₈H₁₁NO Melting Pt: 57 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008137 CAS Number: 7568-93-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...