2-Aminobenzyl alcohol ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

A151122-100G A151122-25G A151122-500G A151122-5G
ALA151122-100GEA 113.91 USD
ALA151122-100G ALA151122-25G ALA151122-500G ALA151122-5G
2-Aminobenzyl alcohol ≥98% (by GC)
2-Aminobenzyl alcohol

2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines.It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde.2-Aminobenzyl alcohol was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 162°C
Melting Pt: 81-83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71c0300c768f899479d90b724e03edc84582fed6&vid=71c0300c768f899479d90b724e03edc84582fed6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR