1-Aminobutan-2-ol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 1-Amino-2-butanol, Monobutanolamine, 1-Aminobutan-2-ol

A151121-5ML A151121-1ML A151121-25ML
ALA151121-5MLEA 72.11 USD
ALA151121-5ML ALA151121-1ML ALA151121-25ML
1-Aminobutan-2-ol ≥98%
1-Aminobutan-2-ol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 169°C
Density: 0.94 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00059196
CAS Number: 13552-21-1
UN: 2735
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7247b9ff18999b47ab98d38a23efd8260c5ab0a&vid=d7247b9ff18999b47ab98d38a23efd8260c5ab0a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR