4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 4-Amino-5-chlorocoumaran-7-carboxylic acid, 4-Amino-5-chloro-2,3-dihydro-1-benzofuran-7-carboxylic acid, 4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic Acid

A151118-10G A151118-1G A151118-250MG A151118-5G
ALA151118-10GEA 2339.76 USD
ALA151118-10G ALA151118-1G ALA151118250MG ALA151118-5G
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid ≥97% (by HPLC, titration analysis)
4-Amino-5-chloro-2,3-dihydrobenzofuran-7-carboxylic acid
Formula: C₉H₈ClNO₃
MW: 213.62 g/mol
Melting Pt: 242 °C
MDL Number: MFCD12923204
CAS Number: 123654-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8a1b98c5d60468c79d5bcf75d44bc756855cc29&vid=c8a1b98c5d60468c79d5bcf75d44bc756855cc29&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR