Aceclofenac ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(2-(2-((2,6-Dichlorophenyl)amino)phenyl)acetoxy)acetic acid, 2-[2-(2,6-Dichloroanilino)phenylacetoxy]acetic Acid

A151117-100G A151117-1G A151117-25G A151117-5G
ALA151117-100GEA 395.11 USD
ALA151117-100G ALA151117-1G ALA151117-25G ALA151117-5G
Aceclofenac ≥98% (by HPLC)
Aceclofenac

Aceclofenac is a non-steroidal anti-inflammatory drug (NSAID) analog of Diclofenac.An anti-inflammatory and analgesic agent

Formula: C₁₆H₁₃Cl₂NO₄
MW: 354.18 g/mol
Boiling Pt: ~486.0°C
Melting Pt: 149-153°C
Density: 1.5 g/cm³
MDL Number: MFCD00864296
CAS Number: 89796-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd31e2d76f7c7b77d80d111b398448120a1a6948&vid=cd31e2d76f7c7b77d80d111b398448120a1a6948&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR