1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
3',4'-Ethylenedioxyacetophenone, 1,4-Benzodioxan-6-yl methyl ketone, 6-Acetyl-1,4-benzodioxane
A151115-1G
A151115-250MG
A151115-25G
A151115-5G
ALA151115-1GEA
12.26
USD
ALA151115-1G
ALA151115250MG
ALA151115-25G
ALA151115-5G
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone ≥98% (by GC)
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Melting Pt: 80-86 °C |
MDL Number:
MFCD00006823 CAS Number: 2879-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...