6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98% (by HPLC, total nitrogen)

Supplier: ALADDIN SCIENTIFIC
Warning

A151113-1G A151113-25G A151113-5G
ALA151113-1GEA 11.31 USD
ALA151113-1G ALA151113-25G ALA151113-5G
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one ≥98% (by HPLC, total nitrogen)
6-Acetyl-2H-1,4-benzoxazin-3(4H)-one

6-Acetyl-2H-1,4-benzoxazin-3(4H)-one, also referred to as 6-acetyl-2H-benzo[b][1,4]oxazin-3(4H)-one, is a useful organic building block.

Formula: C₁₀H₉NO₃
Melting Pt: 194-198°C
MDL Number: MFCD01321313
CAS Number: 26518-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cf1094ed3805c981b5f6df0573c740530106f5b&vid=2cf1094ed3805c981b5f6df0573c740530106f5b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR