2-Amino-3-formylchromone ≥98% (by HPLC, total nitrogen)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-4-oxo-4H-chromene-3-carbaldehyde

A151111-1G A151111-250MG
ALA151111-1GEA 65.46 USD
ALA151111-1G ALA151111250MG
2-Amino-3-formylchromone ≥98% (by HPLC, total nitrogen)
2-Amino-3-formylchromone

Product Description2-Amino-3-formylchromone (2-Amino-4-oxo-4H-1-benzopyran-3-carboxaldehyde) is a benzopyran derivative. It is a bicyclic heterocyclic molecule made up of a benzene ring fused to a heterocyclic pyran ring. It undergoes condensation with (R)-2-amino-2-phenylethanol to afford Schiff base ligand, which forms complexes with Cu(NO3)2 and Zn(NO3)2Product Application2-Amino-3-formylchromone may be used in the preparation of the following:• chiral Schiff base ligands (R)/(S)-2-amino-3-(((1-hydroxypropan-2-yl)imino)methyl)-4H-chromen-4-one• hydrazone derivatives, required for the synthesis of heterocyclic Schiff′s bases having antimicrobial activity[• N-{4-[(6-chloro-4-oxo-4H-chromen-3-ylmethylene)amino]phenyl}-3,5-dimethyl-benzofuran-2-carboxamide• N-{4-[(6-chloro-4-oxo-4H-chromen-3-ylmethylene)amino]phenyl}-1,4-dihydro-chromono [2,3-b]pyrrole-2-carboxamide• 6-chloro-3-{[4-(2-thioxo-3,6-dihydro-2H-1,3,4-thiadiazin-5-ylamino)phenylimino] methyl}- 4-oxo-4H -chromene• 3¬-(2-amino¬-4¬-oxo¬-4H¬-chromen¬-3-¬yl)methylidene)¬-6-¬ ethyl-¬2H-¬pyrano[3,2-¬c]quinolone-¬2,4,5(3H,6H)-trione

Formula: C₁₀H₇NO₃
Melting Pt: 250°C
Storage Temperature: Ambient
MDL Number: MFCD00191735
CAS Number: 61424-76-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f582e96190b6a379d1691529d1db16d72a756d6&vid=9f582e96190b6a379d1691529d1db16d72a756d6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR