Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥99% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
8-Anilino-1-naphthalenesulphonic acid magnesium(II) salt, ANS-Mg, 1,8-ANS magnesium salt, N-Phenyl peri acid magnesium salt, ANSA magnesium salt, Magnesium(II) 8-Anilino-1-naphthalenesulphonate Hydrate, Magnesium 8-(phenylamino)naphthalene-1-sulphonate, Magnesium(II) 8-Anilino-1-naphthalenesulphonate, 8-Anilino-1-naphthalenesulphonic acid Magnesium(II) Salt Hydrate, 8-Anilino-1-naphthalenesulphonic acid hemimagnesium salt hydrate
A151109-1G
A151109-5G
A151109-25G
ALA151109-1GEA
23.66
USD
ALA151109-1G
ALA151109-5G
ALA151109-25G
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate ≥99% (by HPLC)
Magnesium 8-anilinonaphthalene-1-sulphonate hydrate
Formula:
C₃₂H₂₄MgN₂O₆S₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00065353 CAS Number: 18108-68-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...