(Acetylmethylene)triphenylphosphorane ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151107-10G
A151107-5G
ALA151107-10GEA
38.86
USD
ALA151107-10G
ALA151107-5G
(Acetylmethylene)triphenylphosphorane ≥98%
(Acetylmethylene)triphenylphosphorane
Formula:
C₂₁H₁₉OP Melting Pt: 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008774 CAS Number: 1439-36-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...