Allyl trifluoroacetate ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Allyl Trifluoroacetate, ['Trifluoroacetic acid Allyl Ester'], Trifluorethansäure(2-propenyl)ester, Trifluoroacetic allyl ester

A151105-1G A151105-250G A151105-25G A151105-50G A151105-5G
ALA151105-1GEA 9.41 USD
ALA151105-1G ALA151105-250G ALA151105-25G ALA151105-50G ALA151105-5G
Allyl trifluoroacetate ≥95% (by GC)
Allyl trifluoroacetate

Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

Formula: C₅H₅F₃O₂
MW: 154.09
Boiling Pt: 66°C
Flash Pt: -1°C
MDL Number: MFCD00013567
CAS Number: 383-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09170e01981b7fe3acb882107acf2b53bf8a30e2&vid=09170e01981b7fe3acb882107acf2b53bf8a30e2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR