3-Allyloxy-1,2-propanediol ≥99% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151103-25G A151103-100G
ALA151103-25GEA 25.56 USD
ALA151103-25G ALA151103-100G
3-Allyloxy-1,2-propanediol ≥99% (by GC)
3-Allyloxy-1,2-propanediol
Formula: C₆H₁₂O₃
Boiling Pt: 142 °C
Density: 1.068 g/cm³
Flash Pt: 134°C
MDL Number: MFCD00004721
CAS Number: 123-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b5f129c9452b97fbaf76a77d4a25f6fb106405&vid=d8b5f129c9452b97fbaf76a77d4a25f6fb106405&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR