Allyl cyanoacetate ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151102-25G A151102-5G
ALA151102-25GEA 25.56 USD
ALA151102-25G ALA151102-5G
Allyl cyanoacetate ≥97% (by GC)
Allyl cyanoacetate
Formula: C₆H₇NO₂
Boiling Pt: 101°C
Density: 1.074 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00013817
CAS Number: 13361-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f049d3f40f0fa8eff2f0598e0d97ba3255de7dcc&vid=f049d3f40f0fa8eff2f0598e0d97ba3255de7dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR