Allyl benzoate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

A151101-100G A151101-1G A151101-25G A151101-5G
ALA151101-100GEA 180.41 USD
ALA151101-100G ALA151101-1G ALA151101-25G ALA151101-5G
Allyl benzoate ≥98% (by GC)
Allyl benzoate
Formula: C₁₀H₁₀O₂
Boiling Pt: 242°C
Density: 1.054 g/cm³
MDL Number: MFCD00026104
CAS Number: 583-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f712b7e2a0e9b98a157b461373ae1f6b3499c503&vid=f712b7e2a0e9b98a157b461373ae1f6b3499c503&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR