3-(Allyloxy)oxetane ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A151100-1G A151100-250MG
ALA151100-1GEA 73.06 USD
ALA151100-1G ALA151100250MG
3-(Allyloxy)oxetane ≥96%
3-(Allyloxy)oxetane
Formula: C₆H₁₀O₂
Boiling Pt: 64°C
Density: 0.98 g/cm³
MDL Number: MFCD09908098
CAS Number: 6777-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5812b2cdc6c75cfd19285145300ad1a62c69b33e&vid=5812b2cdc6c75cfd19285145300ad1a62c69b33e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR