Allyl nonanoate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151099-25ML A151099-5ML
ALA151099-25MLEA 121.51 USD
ALA151099-25ML ALA151099-5ML
Allyl nonanoate ≥97%
Allyl nonanoate
Formula: C₁₂H₂₂O₂
Boiling Pt: 151°C
Density: 0.88 g/cm³
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038344
CAS Number: 7493-72-3
UN: 1993
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=253a6456e10cd5b5207011331ccb98ff3654e407&vid=253a6456e10cd5b5207011331ccb98ff3654e407&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR