Allyl butyl ether ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Allyl Butyl Ether, Butyl Allyl Ether, 1-(Allyloxy)butane

A151096-100G A151096-5G
ALA151096-100GEA 296.31 USD
ALA151096-100G ALA151096-5G
Allyl butyl ether ≥97% (by GC)
Allyl butyl ether
Formula: C₇H₁₄O
Boiling Pt: 120 °C
Density: 0.783 g/cm³
Flash Pt: 14 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009460
CAS Number: 3739-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f1873476c12d5b7a0aba7b604f3052c1aa3f197&vid=7f1873476c12d5b7a0aba7b604f3052c1aa3f197&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR