Allyl benzyl ether ≥99% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Allyl Benzyl Ether, ((Allyloxy)methyl)benzene
A151095-100ML
A151095-25ML
A151095-5ML
ALA151095100MLEA
243.11
USD
ALA151095100ML
ALA151095-25ML
ALA151095-5ML
Allyl benzyl ether ≥99% (by GC)
Allyl benzyl ether
Formula:
C₁₀H₁₂O Boiling Pt: 204-205 °C Density: 0.959 g/cm³ Flash Pt: 76°C |
MDL Number:
MFCD00078288 CAS Number: 14593-43-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...