Allylcyclohexane ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151092-25ML A151092-5ML
ALA151092-25MLEA 168.06 USD
ALA151092-25ML ALA151092-5ML
Allylcyclohexane ≥97% (by GC)
Allylcyclohexane
Formula: C₉H₁₆
MW: 124.23 g/mol
Boiling Pt: 151°C
Flash Pt: 29°C
MDL Number: MFCD00013769
CAS Number: 2114-42-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4610885d9805ef5eadf01392abde82d484d73cbd&vid=4610885d9805ef5eadf01392abde82d484d73cbd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR