1-Allyl-1H-indole-2,3-dione ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151085-1G A151085-200MG
ALA151085-1GEA 162.36 USD
ALA151085-1G ALA151085200MG
1-Allyl-1H-indole-2,3-dione ≥98% (by GC)
1-Allyl-1H-indole-2,3-dione
Formula: C₁₁H₉NO₂
Boiling Pt: 145°C
Melting Pt: 86 °C
MDL Number: MFCD00224218
CAS Number: 830-74-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1916d625ab52a33c56c96788069898eb77a31c3&vid=a1916d625ab52a33c56c96788069898eb77a31c3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR