2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Azidoethyl 2,3,4,6-Tetra-O-acetyl-β-D-glucopyranoside

A151082-1G A151082-250MG
ALA151082-1GEA 263.06 USD
ALA151082-1G ALA151082250MG
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92% (by HPLC)
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranoside
Formula: C₁₆H₂₃N₃O₁₀
MW: 417.37 g/mol
Melting Pt: 115 °C
Storage Temperature: Refrigerator
CAS Number: 140428-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40fead873735be4397086478a102a760012da668&vid=40fead873735be4397086478a102a760012da668&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR