2-Acetylamino-5-bromo-4-methylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: N-(5-Bromo-4-methylpyridin-2-yl)acetamide, 2-Acetamido-5-bromo-4-picoline, 2-Acetamido-5-bromo-4-methylpyridine

A151079-1G A151079-250MG A151079-25G A151079-5G
ALA151079-1GEA 42.66 USD
ALA151079-1G ALA151079250MG ALA151079-25G ALA151079-5G
2-Acetylamino-5-bromo-4-methylpyridine ≥98%
2-Acetylamino-5-bromo-4-methylpyridine
Formula: C₈H₉BrN₂O
MW: 229.0700 g/mol
Melting Pt: 153 °C
MDL Number: MFCD06798092
CAS Number: 142404-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad4d942531237dde5299fe6a93c1fcf65ad4903&vid=8ad4d942531237dde5299fe6a93c1fcf65ad4903&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR