Amyl laurate ≥97% (by GC)

Supplier: Aladdin Scientific

A151078-25ML A151078-100ML
ALA151078-25MLEA 66.41 USD
ALA151078-25ML ALA151078100ML
Amyl laurate ≥97% (by GC)
Amyl laurate
Formula: C₁₇H₃₄O₂
Density: 0.86 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00059218
CAS Number: 5350-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR