Amyl-n-octanoate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: n-Octanoic acid Amyl Ester,Pentyl n-Octanoate, Pentyl n-Octanoate, Pentyl octanoate, Amyl n-Octanoate, n-Octanoic Acid Amyl Ester

A151073-100ML A151073-25ML
ALA151073100MLEA 94.91 USD
ALA151073100ML ALA151073-25ML
Amyl-n-octanoate ≥98% (by GC)
Amyl-n-octanoate
Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 260°C
Density: 0.86 g/cm³
MDL Number: MFCD00048919
CAS Number: 638-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa1f2a2e0859913f3dc537a08bd21fc3ac05d54c&vid=fa1f2a2e0859913f3dc537a08bd21fc3ac05d54c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR