Acamprosate calcium ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151071-50MG A151071-250MG
ALA151071-50MGEA 61.66 USD
ALA151071-50MG ALA151071250MG
Acamprosate calcium ≥98%
Acamprosate calcium
Formula: C₅H₁₁NO₄S
Melting Pt: 270°C
Storage Temperature: Refrigerator
MDL Number: MFCD00886588
CAS Number: 77337-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db494b1786f5250484c5994d50685ee04f2d4ddf&vid=db494b1786f5250484c5994d50685ee04f2d4ddf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR