Pentyl decanoate ≥93% (by GC)

Supplier: Aladdin Scientific

A151070-25ML A151070-5ML
ALA151070-25MLEA 53.11 USD
ALA151070-25ML ALA151070-5ML
Pentyl decanoate ≥93% (by GC)
Pentyl decanoate
Formula: C₁₅H₃₀O₂
Density: 0.86 g/cm³
Flash Pt: 130 °C
MDL Number: MFCD00059454
CAS Number: 5933-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bb07ed319aa68bbc7f57020a943062da83fe8d0&vid=0bb07ed319aa68bbc7f57020a943062da83fe8d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR