11-Azido-3,6,9-trioxaundecanol ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC

A151069-100MG A151069-1G A151069-500MG
ALA151069100MGEA 65.46 USD
ALA151069100MG ALA151069-1G ALA151069500MG
11-Azido-3,6,9-trioxaundecanol ≥97% (by GC)
11-Azido-3,6,9-trioxaundecanol
Formula: C₈H₁₇N₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD03701128
CAS Number: 86770-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d54949e8cd4dc80a43e09ad2bcb742fa396462e0&vid=d54949e8cd4dc80a43e09ad2bcb742fa396462e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR