4-Acetamidocyclohexanol (cis- and trans- mixture) ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: N-(4-Hydroxycyclohexyl)acetamide(cis- and trans- mixture)

A151058-100G A151058-25G A151058-5G
ALA151058-100GEA 152.86 USD
ALA151058-100G ALA151058-25G ALA151058-5G
4-Acetamidocyclohexanol (cis- and trans- mixture) ≥98% (by GC)
4-Acetamidocyclohexanol (cis- and trans- mixture)
Formula: C₈H₁₅NO₂
Melting Pt: 162°C
MDL Number: MFCD06797038
CAS Number: 23363-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=293f280d5e1ddaafd966394ca8a1fec11488cdb7&vid=293f280d5e1ddaafd966394ca8a1fec11488cdb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR