8-Aminooctan-1-ol ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 8-Amino-1-octanol

A151057-5G A151057-1G A151057-250MG A151057-25G
ALA151057-5GEA 366.61 USD
ALA151057-5G ALA151057-1G ALA151057250MG ALA151057-25G
8-Aminooctan-1-ol ≥95%
8-Aminooctan-1-ol
Formula: C₈H₁₉NO
MW: 145.2400 g/mol
Melting Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD02094315
CAS Number: 19008-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15d859ebd6fa2f01a97b5df0ae2f337aac4e80ab&vid=15d859ebd6fa2f01a97b5df0ae2f337aac4e80ab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR