Azobenzene-4,4'-dicarboxylic acid ≥95% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Azobenzene-4,4'-dicarboxylic Acid, 4,4'-Azodibenzoic acid, 4,4'-Azodibenzoic Acid, 4,4'-(Diazene-1,2-diyl)dibenzoic acid

A151052-10G A151052-1G A151052-250MG
ALA151052-10GEA 655.41 USD
ALA151052-10G ALA151052-1G ALA151052250MG
Azobenzene-4,4'-dicarboxylic acid ≥95% (by HPLC, titration analysis)
Azobenzene-4,4'-dicarboxylic acid
Formula: C₁₄H₁₀N₂O₄
MW: 270.24 g/mol
MDL Number: MFCD00496621
CAS Number: 586-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26d180292b93148e3c2a33efcd1ca83fbd9d5df3&vid=26d180292b93148e3c2a33efcd1ca83fbd9d5df3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR