4-Aminotriphenylamine ≥95% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N,N-Diphenyl-1,4-phenylenediamine, 4-Amino-N,N-diphenylaniline

A151051-1G A151051-250MG A151051-50MG
ALA151051-1GEA 391.31 USD
ALA151051-1G ALA151051250MG ALA151051-50MG
4-Aminotriphenylamine ≥95% (by HPLC)
4-Aminotriphenylamine
Formula: C₁₈H₁₆N₂
Boiling Pt: 270°C
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00145130
CAS Number: 2350-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR