2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: N-([1,1'-Biphenyl]-4-yl)-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine

A151050-1G A151050-250MG
ALA151050-1GEA 35.06 USD
ALA151050-1G ALA151050250MG
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene ≥98% (by HPLC)
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene
Formula: C₃₃H₂₆BrN
MW: 516.48 g/mol
Melting Pt: 159.0 to 163.0 °C
MDL Number: MFCD20040455
CAS Number: 1246562-40-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b85c6beeb6257fdd3af7ff2ee805a3daac15be2&vid=5b85c6beeb6257fdd3af7ff2ee805a3daac15be2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR