2-(Aminomethyl)-1,3-dioxolane ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151048-100MG
A151048-1G
A151048-250MG
A151048-5G
ALA151048100MGEA
22.71
USD
ALA151048100MG
ALA151048-1G
ALA151048250MG
ALA151048-5G
2-(Aminomethyl)-1,3-dioxolane ≥98%
2-(Aminomethyl)-1,3-dioxolane
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142862 CAS Number: 4388-97-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...