Allyl ethyl carbonate ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151039-1ML
A151039-25ML
A151039-5ML
ALA151039-1MLEA
22.71
USD
ALA151039-1ML
ALA151039-25ML
ALA151039-5ML
Allyl ethyl carbonate ≥97%
Allyl ethyl carbonate
Formula:
C₆H₁₀O₃ Boiling Pt: 149°C Density: 0.99 g/cm³ Flash Pt: 46 °C |
MDL Number:
MFCD00049113 CAS Number: 1469-70-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...