Allyl phenyl carbonate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151037-1G A151037-5G
ALA151037-1GEA 13.21 USD
ALA151037-1G ALA151037-5G
Allyl phenyl carbonate ≥97%
Allyl phenyl carbonate
Formula: C₁₀H₁₀O₃
Boiling Pt: 78°C
Density: 1.11 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD07784342
CAS Number: 16308-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=593d05fa0dc600a478bf24566d16ff8e4749af4d&vid=593d05fa0dc600a478bf24566d16ff8e4749af4d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR