Allyl-2,2,2-Trichloroacetimidate ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151034-1G
A151034-25G
A151034-5G
ALA151034-1GEA
28.41
USD
ALA151034-1G
ALA151034-25G
ALA151034-5G
Allyl-2,2,2-Trichloroacetimidate ≥98% (by GC)
Allyl-2,2,2-Trichloroacetimidate
Formula:
C₅H₆Cl₃NO Boiling Pt: 74°C Density: 1.34 g/cm³ Flash Pt: 74°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00190710 CAS Number: 51479-73-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...