Allyl-ɑ-D-galactopyranoside ≥96% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Ally a-D-galactopyranoside, Allyl α-D-Galactopyranoside, 2-Propen-1-yl α-D-Galactopyranoside, α-D-Galactose Monoallyl Ether

A151031-1G A151031-250MG A151031-5G
ALA151031-1GEA 52.16 USD
ALA151031-1G ALA151031250MG ALA151031-5G
Allyl-ɑ-D-galactopyranoside ≥96% (by HPLC)
Allyl-alpha-D-galactopyranoside
Formula: C₉H₁₆O₆
Melting Pt: 138-141°C
Storage Temperature: Freezer
MDL Number: MFCD01320363
CAS Number: 48149-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca07dca8a0dd37acd6e4c9933c0818c1e3c0d4d8&vid=ca07dca8a0dd37acd6e4c9933c0818c1e3c0d4d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR