Argatroban monohydrate ≥99% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

A151030-100MG A151030-250MG A151030-25MG A151030-5MG
ALA151030100MGEA 666.81 USD
ALA151030100MG ALA151030250MG ALA151030-25MG ALA151030-5MG
Argatroban monohydrate ≥99% (by HPLC)
Argatroban monohydrate
Formula: C₂₃H₃₆N₆O₅S·H₂O
Melting Pt: 180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00895735
CAS Number: 141396-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa74b09b0081ae76d7488419d4acbc67fa3505f8&vid=fa74b09b0081ae76d7488419d4acbc67fa3505f8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR