4-Aminomorpholine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
N-Aminomorpholine
A151029-25ML
A151029-5ML
A151029-100ML
ALA151029-25MLEA
75.91
USD
ALA151029-25ML
ALA151029-5ML
ALA151029100ML
4-Aminomorpholine ≥98% (by GC)
4-Aminomorpholine
Formula:
C₄H₁₀N₂O Boiling Pt: 168 °C Density: 1.059 g/cm³ Flash Pt: 58°C Storage Temperature: Ambient |
MDL Number:
MFCD00006174 CAS Number: 4319-49-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...