4-Allylmorpholine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151028-1ML A151028-5ML
ALA151028-1MLEA 13.21 USD
ALA151028-1ML ALA151028-5ML
4-Allylmorpholine ≥97% (by GC)
4-Allylmorpholine
Formula: C₇H₁₃NO
Boiling Pt: 158°C
Density: 0.93 g/cm³
Flash Pt: 43 °C
MDL Number: MFCD00038034
CAS Number: 696-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddfb9920e66f3097ae1d16fca14abaaa9bd27e18&vid=ddfb9920e66f3097ae1d16fca14abaaa9bd27e18&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR