4-Acetamidobenzaldehyde ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: N-(4-Formylphenyl)acetamide, 4'-Formylacetanilide

A151024-25G A151024-100G
ALA151024-25GEA 94.91 USD
ALA151024-25G ALA151024-100G
4-Acetamidobenzaldehyde ≥98% (by GC)
4-Acetamidobenzaldehyde
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 154-158°C
Storage Temperature: Ambient
MDL Number: MFCD00003380
CAS Number: 122-85-0
EINECS: 204-579-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d6392e7c33dde143953f79ce71fc1ba0ff7a01f&vid=4d6392e7c33dde143953f79ce71fc1ba0ff7a01f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR