anti-(1R)-(+)-Camphorquinone 3-oxime ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: anti-(1R)-(+)-Camphorquinone 3-Oxime, (1R,E)-(+)-2,3-Bornanedione 3-oxime,anti-(1R)-(+)-Camphorquinone 3-oxime, anti-(1R)-(+)-2,3-Bornanedione 3-Oxime

A151019-1G A151019-250MG
ALA151019-1GEA 117.71 USD
ALA151019-1G ALA151019250MG
anti-(1R)-(+)-Camphorquinone 3-oxime ≥95%
anti-(1R)-(+)-Camphorquinone 3-oxime

Product Application:(1R,E)-(+)-Camphorquinone 3-oxime can be used to prepare chiral camphor-oxazoline auxiliary, which is used to blend diastereoselectivity and isomer separability for the efficient synthesis of bicuculline, egenine, corlumine, and corytensine.

Formula: C₁₀H₁₅NO₂
MW: 181.24 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00074848
CAS Number: 31571-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cde640637a1d3ab914e7836d2fa51e3dc31162d&vid=5cde640637a1d3ab914e7836d2fa51e3dc31162d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR