2-Acetyl-5-chlorothiophene ≥99% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151017-100G A151017-25G A151017-5G
ALA151017-100GEA 254.51 USD
ALA151017-100G ALA151017-25G ALA151017-5G
2-Acetyl-5-chlorothiophene ≥99% (by GC)
2-Acetyl-5-chlorothiophene
Formula: C₆H₅ClOS
Boiling Pt: 118°C
Melting Pt: 46 °C
Flash Pt: 108 °C
MDL Number: MFCD00005444
CAS Number: 6310-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR